missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cathepsin K Protein

A DNA sequence encoding the Homo sapiens (Human) Cathepsin K, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5889-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15938489

  • £1265.00 / 500µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000387.1
Cathepsin K Protein
Research Use Only
CTSK
Recombinant Protein
Yeast
APDSVDYRKK GYVTPVKNQG QCGSCWAFSS VGALEGQLKK KTGKLLNLSP QNLVDCVSEN DGCGGGYMTN AFQYVQKNRG IDSEDAYPYV GQEESCMYNP TGKAAKCRGY REIPEGNEKA LKRAVARVGP VSVAIDASLT SFQFYSKGVY YDESCNSDNL NHAVLAVGYG IQKGNKHWII KNSWGENWGN KGYILMARNK NNACGIANLA SFPKM
Greater than 90% as determined by SDS-PAGE.
1513
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Product Certifications
Ofertas especiales

Ofertas especiales