missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AKT2 / Protein
A DNA sequence encoding the Homo sapiens (Human) AKT2 /, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001317440.1 |
---|---|
Gene ID (Entrez) | 208 |
Name | AKT2 / Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15916639
|
enQuireBio™
QP5641-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936639
|
enQuireBio™
QP5641-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926639
|
enQuireBio™
QP5641-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15946639
|
enQuireBio™
QP5641-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15956639
|
enQuireBio™
QP5641-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15966639
|
enQuireBio™
QP5641-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001317440.1 | |
AKT2 / Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
208 | |
Research Use Only | |
AKT2 | |
Recombinant Protein | |
E. coli | |
NEVSVIKEGW LHKRGEYIKT WRPRYFLLKS DGSFIGYKER PEAPDQTLPP LNNFSVAECQ LMKTERPRPN TFVIRCLQWT TVIERTFHVD SPDEREEWMR AIQMVANSLK QRAPGEDPMD YKCGSPSDSS TTEEMEVAVS KARAKVTMND FDYLKLLGKG TFGKVILVRE KATGRYYAMK ILRKEVIIAK DEVAHTVTES RVLQNTRHPF LTALKYAFQT HDRLCFVMEY ANGGELFFHL SRERVFTEER ARFYGAEIVS ALEYLHSRDV VYRDIKLENL MLDKDGHIKI TDFGLCKEGI SDGATMKTFC GTPEYLAPEV LEDNDYGRAV DWWGLGVVMY EMMCGRLPFY NQDHERLFEL ILMEEIRFPR TLSPEAKSLL AGLLKKDPKQ RLGGGPSDAK EVMEHRFFLS INWQDVVQKK LLPPFKPQVT SEVDTRYFDD EFTAQSITIT PPDRYDSLGL LELDQRTHFP QFSYSASIRE | |
Greater than 90% as determined by SDS-PAGE. |