missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AKT2 / Protein

A DNA sequence encoding the Homo sapiens (Human) AKT2 /, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5641-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936639

  • £1735.00 / 1mg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001317440.1
AKT2 / Protein
Research Use Only
AKT2
Recombinant Protein
E. coli
NEVSVIKEGW LHKRGEYIKT WRPRYFLLKS DGSFIGYKER PEAPDQTLPP LNNFSVAECQ LMKTERPRPN TFVIRCLQWT TVIERTFHVD SPDEREEWMR AIQMVANSLK QRAPGEDPMD YKCGSPSDSS TTEEMEVAVS KARAKVTMND FDYLKLLGKG TFGKVILVRE KATGRYYAMK ILRKEVIIAK DEVAHTVTES RVLQNTRHPF LTALKYAFQT HDRLCFVMEY ANGGELFFHL SRERVFTEER ARFYGAEIVS ALEYLHSRDV VYRDIKLENL MLDKDGHIKI TDFGLCKEGI SDGATMKTFC GTPEYLAPEV LEDNDYGRAV DWWGLGVVMY EMMCGRLPFY NQDHERLFEL ILMEEIRFPR TLSPEAKSLL AGLLKKDPKQ RLGGGPSDAK EVMEHRFFLS INWQDVVQKK LLPPFKPQVT SEVDTRYFDD EFTAQSITIT PPDRYDSLGL LELDQRTHFP QFSYSASIRE
Greater than 90% as determined by SDS-PAGE.
208
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers