missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AKT2 / Protein
A DNA sequence encoding the Homo sapiens (Human) AKT2 /, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5641-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001317440.1 | |
AKT2 / Protein | |
Research Use Only | |
AKT2 | |
Recombinant Protein | |
E. coli | |
NEVSVIKEGW LHKRGEYIKT WRPRYFLLKS DGSFIGYKER PEAPDQTLPP LNNFSVAECQ LMKTERPRPN TFVIRCLQWT TVIERTFHVD SPDEREEWMR AIQMVANSLK QRAPGEDPMD YKCGSPSDSS TTEEMEVAVS KARAKVTMND FDYLKLLGKG TFGKVILVRE KATGRYYAMK ILRKEVIIAK DEVAHTVTES RVLQNTRHPF LTALKYAFQT HDRLCFVMEY ANGGELFFHL SRERVFTEER ARFYGAEIVS ALEYLHSRDV VYRDIKLENL MLDKDGHIKI TDFGLCKEGI SDGATMKTFC GTPEYLAPEV LEDNDYGRAV DWWGLGVVMY EMMCGRLPFY NQDHERLFEL ILMEEIRFPR TLSPEAKSLL AGLLKKDPKQ RLGGGPSDAK EVMEHRFFLS INWQDVVQKK LLPPFKPQVT SEVDTRYFDD EFTAQSITIT PPDRYDSLGL LELDQRTHFP QFSYSASIRE | |
Greater than 90% as determined by SDS-PAGE. |
208 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |