missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™175mm Diameter Glass Büchner Filter Funnel
Manufactured from chemically resistant Pyrex™ glass. Pyrex™ 175mm Diameter Glass Büchner Filter Funnel with integral sintered glass disc is highly resistant to most corrosive reagents and eliminates the need for filter papers.
£309.00 - £626.00
Specifications
Capacity (Metric) | 4000 mL |
---|---|
Diameter (Metric) Stem | 30mm |
Material | Pyrex Glass, Borosilicate Glass |
Height (Metric) | 390 mm |
Diameter (Metric) Disc | 175mm |
Product Code | Brand | Porosity | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Pore Size | Price | Quantity & Availability | ||||
11368004
|
Pyrex
3722/02M |
1 | 100 to 160μm |
£309.00
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
11398004
|
Pyrex
3722/08M |
4 | 10 to 16μm |
£403.00
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
11388004
|
Pyrex
3722/06M |
3 | 16 to 40μm |
£626.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
Description
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid - Choice of disc diameters and porosities
Specifications
4000 mL | |
Pyrex Glass, Borosilicate Glass | |
175mm | |
150mm |
30mm | |
390 mm | |
Integrated filter plate sintered Glass disc |