Learn More
Pyrex™175mm Diameter Glass Büchner Filter Funnel
Manufactured from chemically resistant Pyrex™ glass. Pyrex™ 175mm Diameter Glass Büchner Filter Funnel with integral sintered glass disc is highly resistant to most corrosive reagents and eliminates the need for filter papers.
Brand: Pyrex 3722/06M
337.71 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : A0
Additional Details : Weight : 1.65000kg
Description
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid - Choice of disc diameters and porosities
Specifications
Buchner filter funnel | |
30mm | |
390 mm | |
175mm | |
150mm | |
16 to 40μm |
4000 mL | |
Pyrex Glass, Borosilicate Glass | |
For easy filtering solutions | |
Integrated filter plate sintered Glass disc | |
3 |