missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™175mm Diameter Glass Büchner Filter Funnel
Description
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid - Choice of disc diameters and porosities
Specifications
Specifications
| Capacity (Metric) | 4000 mL |
| Diameter (Metric) Stem | 30mm |
| Material | Borosilicate Glass |
| Height (Metric) | 390 mm |
| For Use With (Application) | Fine precipitate analytical filtration |
| Diameter (Metric) Disc | 175mm |
| Includes | Integrated filter plate sintered Glass disc |
| Length (Metric) Stem | 150mm |
| Porosity | 4 |
| Pore Size | 10 to 16μm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?