Learn More
Agilent BioTek Imaging Objectives for Lionheart FX Automated Live Cell Imager
For use with Lionheart FX Automated Live Cell Imager.
Brand:  Agilent BioTek 1450538
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 16542152
  • £17045.00 / Each
Estimated Shipment: 22-05-2024
to see stock.
Description
Description
  • Includes oil and absorbing ring
Specifications
Specifications
60x Oil Objective Olympus, 1.42 NA
Lionheart FX Automated Live Cell Imager