Learn More
Agilent BioTek Imaging Objectives for Lionheart FX Automated Live Cell Imager
For use with Lionheart FX Automated Live Cell Imager.
£17045.00
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
16542152
|
Agilent BioTek
1450538 |
60x Oil Objective Olympus, 1.42 NA |
£17045.00
Each |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16552152
|
Agilent BioTek
1450539 |
100X Oil Objective Olympus, 1.40 NA |
£17045.00
Each |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Includes oil and absorbing ring
Specifications
Lionheart FX Automated Live Cell Imager |