missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DSS (disuccinimidyl suberate), No-Weigh™ Format
Crosslink amines to amines with disuccinimidyl suberate composed of NHS-ester groups with a membrane-permeable, mid-length spacer for intracellular protein crosslinking.
£76.25 - £217.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11831255
|
Thermo Scientific™
21658 |
8 x 2 mg | |||||||
15971252
|
Thermo Scientific™
A39267 |
10 x 2 mg |
£89.50
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10138353
|
Thermo Scientific™
21655 |
50 mg |
£80.25
50mg |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10540434
|
Thermo Scientific™
21555 |
1 g |
£217.00
1g |
Estimated Shipment: 01-10-2024
Log in to see stock. |
|||||
Description
Thermo Scientific™ Pierce DSS, also called disuccinimidyl suberate, is noncleavable and membrane permeable crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm.
Highlights:
- Chemical crosslinking of intracellular proteins prior to cell lysis and immunoprecipitation
- “Fix” protein interactions to allow identification of weak or transient protein interactions
- Protein crosslinking to create bioconjugates via single-step reactions
- Crosslink oriented antibodies to Protein A or G agarose
- Immobilize proteins onto amine-coated surfaces
Specifications
No | |
No |
For Research Use Only. Not for use in diagnostic procedures.