missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DSS (disuccinimidyl suberate), No-Weigh™ Format

Product Code. 11831255 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
8 x 2 mg
10 x 2 mg
1 g
Unit Size:
1g
50mg
Pack of 10
Pack of 8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11831255 8 x 2 mg Pack of 8
15971252 10 x 2 mg Pack of 10
10138353 50 mg 50mg
10540434 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11831255 Supplier Thermo Scientific™ Supplier No. 21658

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Crosslink amines to amines with disuccinimidyl suberate composed of NHS-ester groups with a membrane-permeable, mid-length spacer for intracellular protein crosslinking.

Thermo Scientific™ Pierce DSS, also called disuccinimidyl suberate, is noncleavable and membrane permeable crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm.

Highlights:

  • Chemical crosslinking of intracellular proteins prior to cell lysis and immunoprecipitation
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Crosslink oriented antibodies to Protein A or G agarose
  • Immobilize proteins onto amine-coated surfaces
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-to-Amine
Cleavable No
Description Pierce DSS
Formulation Disuccinimidyl suberate; powder solid
Membrane Permeable Yes
Molecular Weight (g/mol) 368.34
PEGylated No
Sufficient For 43mL of typical 1X (1mM) reaction solution (total), or 8 separate uses
Spacer Arm Length 11.4Å
Quantity 8 x 2 mg
Solubility Not soluble in water
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.