missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p300/CBP (CREB-Binding Protein) Ab-1 Mouse Monoclonal Antibody, Epredia™
Achieve accurate, reproducible results with the Epredia™ p300/CBP (CREB-Binding Protein) Ab-1, Mouse Monoclonal Antibody.
£118.32 - £561.00
Specifications
Antigen | p300/CBP (CREB-Binding Protein) Ab-1 |
---|---|
Clone | NM11 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12671677
|
Epredia
MS-586-P1 |
Concentrated | Unconjugated | 500 μL |
£409.00
500µL |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12641677
|
Epredia
MS-586-B1 |
Concentrated | Biotin | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12661677
|
Epredia
MS-586-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12681677
|
Epredia
MS-586-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12691677
|
Epredia
MS-586-PABX |
Concentrated | Unconjugated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12631677
|
Epredia
MS-586-B0 |
Concentrated | Biotin | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12651677
|
Epredia
MS-586-P |
Concentrated | Unconjugated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12621687
|
Epredia
MS-586-R7 |
Prediluted | Unconjugated | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
p300 is a transcriptional adapter protein which is characterized by three cysteine- and histidine-rich regions and its C-terminus specifically binds the adenovirus E1A protein. p300, and p300-associated proteins, are components of TATA-binding protein (TBP) complexes. Protein kinase A mediated CREB phosphorylation results in the binding of CREB to a 265kDa nuclear protein designated CBP (for CREB-binding protein). CBP and p300 are homologous to each other.Host Species: Mouse
Clone: NM11
Isotype: IgG2b/κ
Species Reactivity: Human, Monkey, Mink, Mouse, and Rat. Others not known.
Epitope: aa 2071-2091
Immunogen: E1A affinity purified p300 protein
Molecular Weight: 300kDa
Positive Control: Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
p300/CBP (CREB-Binding Protein) Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Mouse | |
Transcription Regulation | |
E1A affinity purified p300 protein | |
Primary |
NM11 | |
Monoclonal | |
RUO | |
Human, Mink, Mouse, Non-human Primate, Rat | |
IgG2b κ |