Learn More
p300/CBP (CREB-Binding Protein) Ab-1 Mouse Monoclonal Antibody, Epredia™
Achieve accurate, reproducible results with the Epredia™ p300/CBP (CREB-Binding Protein) Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-586-P1ABX
Code : LD
Additional Details : Weight : 1.00000kg
Description
p300 is a transcriptional adapter protein which is characterized by three cysteine- and histidine-rich regions and its C-terminus specifically binds the adenovirus E1A protein. p300, and p300-associated proteins, are components of TATA-binding protein (TBP) complexes. Protein kinase A mediated CREB phosphorylation results in the binding of CREB to a 265kDa nuclear protein designated CBP (for CREB-binding protein). CBP and p300 are homologous to each other.Host Species: Mouse
Clone: NM11
Isotype: IgG2b/κ
Species Reactivity: Human, Monkey, Mink, Mouse, and Rat. Others not known.
Epitope: aa 2071-2091
Immunogen: E1A affinity purified p300 protein
Molecular Weight: 300kDa
Positive Control: Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
p300/CBP (CREB-Binding Protein) Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Transcription Regulation | |
Human, Mink, Mouse, Non-human Primate, Rat | |
IgG2b κ |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
NM11 | |
Unconjugated | |
E1A affinity purified p300 protein | |
RUO | |
Primary | |
Concentrated |