missing translation for 'onlineSavingsMsg'
Learn More

p300/CBP (CREB-Binding Protein) Ab-1 Mouse Monoclonal Antibody, Epredia™

Achieve accurate, reproducible results with the Epredia™ p300/CBP (CREB-Binding Protein) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-586-P1

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12671677

  • £409.00 / 500µL
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

p300 is a transcriptional adapter protein which is characterized by three cysteine- and histidine-rich regions and its C-terminus specifically binds the adenovirus E1A protein. p300, and p300-associated proteins, are components of TATA-binding protein (TBP) complexes. Protein kinase A mediated CREB phosphorylation results in the binding of CREB to a 265kDa nuclear protein designated CBP (for CREB-binding protein). CBP and p300 are homologous to each other.

Host Species: Mouse

Clone: NM11

Isotype: IgG2b/κ

Species Reactivity: Human, Monkey, Mink, Mouse, and Rat. Others not known.

Epitope: aa 2071-2091

Immunogen: E1A affinity purified p300 protein

Molecular Weight: 300kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p300/CBP (CREB-Binding Protein) Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Transcription Regulation
Human, Mink, Mouse, Non-human Primate, Rat
IgG2b κ
Immunofluorescence, Immunohistochemistry (Paraffin)
NM11
Unconjugated
E1A affinity purified p300 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers