missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mucin 2 (MUC2) Ab-2 Mouse Monoclonal Antibody, Epredia™
Deliver accurate, reproducible results with the Epredia™ Mucin 2 (MUC2) Ab-2, Mouse Monoclonal Antibody.
£141.00 - £576.30
Specifications
Antigen | Mucin 2 (MUC2) Ab-2 |
---|---|
Clone | M53 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
12688686
|
Epredia
MS-1037-P0 |
Concentrated | 200μg/mL | 100 μL |
£141.00
100µL |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12698686
|
Epredia
MS-1037-P1 |
Concentrated | 200μg/mL | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12678686
|
Epredia
MS-1037-P |
Concentrated | 200μg/mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12618696
|
Epredia
MS-1037-P1ABX |
Concentrated | 1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12608696
|
Epredia
MS-1037-PABX |
Concentrated | 1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12648696
|
Epredia
MS-1037-R7 |
Prediluted | — | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12658696
|
Epredia
MS-1037-RQ |
Ready-to-Use | — | 12 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.Host Species: Mouse
Clone: M53
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein encoding five tandem repeats of the human MUC2 apomucin
Molecular Weight: 520kDa
Positive Control: Small intestine
Cellular Localization: Cytoplasmic and cell surface
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Mucin 2 (MUC2) Ab-2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Recombinant protein encoding 5 tandem repeats of the human MUC2 apomucin |
M53 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Primary |