missing translation for 'onlineSavingsMsg'
Learn More

Mucin 2 (MUC2) Ab-2 Mouse Monoclonal Antibody, Epredia™

Deliver accurate, reproducible results with the Epredia™ Mucin 2 (MUC2) Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia MS-1037-P0

 View more versions of this product

Product Code. 12688686

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.

Host Species: Mouse

Clone: M53

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant protein encoding five tandem repeats of the human MUC2 apomucin

Molecular Weight: 520kDa

Positive Control: Small intestine

Cellular Localization: Cytoplasmic and cell surface

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mucin 2 (MUC2) Ab-2
Monoclonal
200μg/mL
Mouse
100 μL
Cancer and Tumor Biology
Human
IgG2a κ
Immunohistochemistry (Paraffin)
M53
Unconjugated
Recombinant protein encoding 5 tandem repeats of the human MUC2 apomucin
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.