Learn More
Mucin 2 (MUC2) Ab-2 Mouse Monoclonal Antibody, Epredia™
Deliver accurate, reproducible results with the Epredia™ Mucin 2 (MUC2) Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia MS-1037-RQ
Code : LD
Additional Details : Weight : 0.00190kg
Description
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.Host Species: Mouse
Clone: M53
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein encoding five tandem repeats of the human MUC2 apomucin
Molecular Weight: 520kDa
Positive Control: Small intestine
Cellular Localization: Cytoplasmic and cell surface
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Mucin 2 (MUC2) Ab-2 | |
Monoclonal | |
— | |
Mouse | |
12 mL | |
Cancer and Tumor Biology | |
Ready-to-Use |
Immunohistochemistry (Paraffin) | |
M53 | |
Unconjugated | |
Recombinant protein encoding 5 tandem repeats of the human MUC2 apomucin | |
IVD | |
Primary |