missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Magnetic Filters Funnel, Non-sterile
Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes.
£300.00 - £420.02
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17124281
|
Cytiva (formerly Pall Lab)
4238 |
500 mL |
£420.02
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17184281
|
Cytiva (formerly Pall Lab)
4247 |
150 mL |
£300.00
Each |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
17144281
|
Cytiva (formerly Pall Lab)
4242 |
300 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Not approved for use in medical, clinical, surgical or other patient applications
Specifications
Polyphenylsulfone | |
47 mm |