missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Magnetic Filters Funnel, Non-sterile

Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes.

Brand:  Cytiva (formerly Pall Lab) 4247

Additional Details : Weight : 0.24700kg

 View more versions of this product

Product Code. 17184281

  • £300.00 / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket

Description

Description

  • Not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

150 mL
Polyphenylsulfone
EFA: 9.6 cm2
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers