missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Magnetic Filters Funnel, Non-sterile

Product Code. 17124281
Change view
Click to view available options
Capacity (Metric):
150 mL
300 mL
500 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17124281 500 mL Each
17144281 300 mL Each
17184281 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17124281 Supplier Cytiva Supplier No. 4238
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes.

  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Material PPSF
Autoclavable Autoclavable
Description EFA: 13.1 cm2
Diameter (Metric) Filter 47 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.