Learn More
Lidocaine hydrochloride monohydrate, 98%, Thermo Scientific™
£23.65 - £52.20
Chemical Identifiers
| CAS | 6108-05-0 |
|---|---|
| Molecular Formula | C14H25ClN2O2 |
| Molecular Weight (g/mol) | 288.82 |
| MDL Number | MFCD00150329 |
| InChI Key | YECIFGHRMFEPJK-UHFFFAOYSA-N |
| Synonym | lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate |
| PubChem CID | 16219577 |
| ChEBI | CHEBI:60791 |
| IUPAC Name | {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride |
| SMILES | O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15415309
|
Thermo Scientific Alfa Aesar
J63035.06 |
5g |
£26.30 £23.65 / 5g Save £2.65 10% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15425309
|
Thermo Scientific Alfa Aesar
J63035.14 |
25g |
£52.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is also used as a fast voltage-gated sodium channel blocker.
Solubility
Soluble in water (50 mg/ml), chloroform, ethanol, and benzene. Insoluble in diethyl ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Do not store above 40 degree C (104 0 F). (preferably store at temperature between 15degree C and 30 degree C).
Chemical Identifiers
| 6108-05-0 | |
| 288.82 | |
| YECIFGHRMFEPJK-UHFFFAOYSA-N | |
| 16219577 | |
| {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride |
| C14H25ClN2O2 | |
| MFCD00150329 | |
| lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate | |
| CHEBI:60791 | |
| O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C |
Specifications
| 6108-05-0 | |
| MFCD00150329 | |
| 14,5482 | |
| Soluble in water (50mg/ml),chloroform,ethanol,and benzene. Insoluble in diethyl ether. | |
| O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C | |
| 288.82 | |
| CHEBI:60791 | |
| 98% |
| C14H25ClN2O2 | |
| UN2811 | |
| lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate | |
| YECIFGHRMFEPJK-UHFFFAOYSA-N | |
| {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride | |
| 16219577 | |
| 288.82 (270.80 Anhydrous) | |
| Lidocaine hydrochloride monohydrate |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : AN7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.