missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine hydrochloride monohydrate, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J63035.06

Quantity 5g

 View more versions of this product

Product Code. 15415309

  • ONLINE EXCLUSIVE PRICE
    £26.30
    £23.65 / 5g
    Save £2.65
    10% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is also used as a fast voltage-gated sodium channel blocker.

Solubility
Soluble in water (50 mg/ml), chloroform, ethanol, and benzene. Insoluble in diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Do not store above 40 degree C (104 0 F). (preferably store at temperature between 15degree C and 30 degree C).
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

6108-05-0
288.82
YECIFGHRMFEPJK-UHFFFAOYSA-N
16219577
{[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride
C14H25ClN2O2
MFCD00150329
lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate
CHEBI:60791
O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C

Specifications

6108-05-0
MFCD00150329
UN2811
lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate
YECIFGHRMFEPJK-UHFFFAOYSA-N
{[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride
16219577
288.82 (270.80 Anhydrous)
Lidocaine hydrochloride monohydrate
C14H25ClN2O2
5g
14,5482
Soluble in water (50mg/ml),chloroform,ethanol,and benzene. Insoluble in diethyl ether.
O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C
288.82
CHEBI:60791
98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.