accessibility menu, dialog, popup

UserName

H2OClH3CNHH3CONHCH3H3C

CAS RN 6108-05-0

IUPAC Name: {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride
Synonyms: xylocard lidocaine hydrochloride monohydrate
Molecular Weight (g/mol): 288.82
Molecular Formula: C14H25ClN2O2
InChi Key: YECIFGHRMFEPJK-UHFFFAOYSA-N
SMILES: O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C
Molecular Weight (g/mol) 
  • (4)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (3)
  • (4)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)