Learn More
Lidocaine hydrochloride monohydrate, 98%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J63035.14
| Quantity | 25g |
|---|
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is also used as a fast voltage-gated sodium channel blocker.
Solubility
Soluble in water (50 mg/ml), chloroform, ethanol, and benzene. Insoluble in diethyl ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Do not store above 40 degree C (104 0 F). (preferably store at temperature between 15degree C and 30 degree C).
Chemical Identifiers
| 6108-05-0 | |
| 288.82 | |
| YECIFGHRMFEPJK-UHFFFAOYSA-N | |
| 16219577 | |
| {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride |
| C14H25ClN2O2 | |
| MFCD00150329 | |
| lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate | |
| CHEBI:60791 | |
| O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C |
Specifications
| 6108-05-0 | |
| MFCD00150329 | |
| UN2811 | |
| lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate | |
| YECIFGHRMFEPJK-UHFFFAOYSA-N | |
| {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride | |
| 16219577 | |
| 288.82 (270.80 Anhydrous) | |
| Lidocaine hydrochloride monohydrate |
| C14H25ClN2O2 | |
| 25g | |
| 14,5482 | |
| Soluble in water (50mg/ml),chloroform,ethanol,and benzene. Insoluble in diethyl ether. | |
| O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C | |
| 288.82 | |
| CHEBI:60791 | |
| 98% |
Safety and Handling
- Lidocaine hydrochloride monohydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
- MIXTURE LIST-Contain: Lidocaine hydrochloride monohydrate
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.