Learn More
Vascular Endothelial Growth Factor (VEGF) Ab-7 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Vascular Endothelial Growth Factor (VEGF) Ab-7, Mouse Monoclonal Antibody.
£131.58 - £653.00
Specifications
Antigen | Vascular Endothelial Growth Factor (VEGF) Ab-7 |
---|---|
Clone | VG1 |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12649876
|
Epredia
MS-1467-P1 |
Concentrated | 500 μL |
£418.00
500µL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12629876
|
Epredia
MS-1467-P |
Concentrated | 1 mL |
£653.00
1mL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12669876
|
Epredia
MS-1467-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12639876
|
Epredia
MS-1467-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12659876
|
Epredia
MS-1467-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12699876
|
Epredia
MS-1467-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
VEGF which is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.Host Species: Mouse
Clone: VG1
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant VEGF protein
Molecular Weight: 19-22kDa (reduced)
Positive Control: VEGF165 recombinant protein, Angiosarcoma Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-7 does not neutralize the bioactivity of VEGF. Recommended for: Note: USA: RUO; Int'l: RUO
Specifications
Vascular Endothelial Growth Factor (VEGF) Ab-7 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RUO | |
Human | |
IgG2a κ |
VG1 | |
Monoclonal | |
Mouse | |
Angiogenesis | |
Recombinant VEGF protein | |
Primary |