Learn More
Vascular Endothelial Growth Factor (VEGF) Ab-7 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Vascular Endothelial Growth Factor (VEGF) Ab-7, Mouse Monoclonal Antibody.
Brand: Epredia MS-1467-P
Code : LD
Additional Details : Weight : 0.00250kg
Description
VEGF which is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.Host Species: Mouse
Clone: VG1
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant VEGF protein
Molecular Weight: 19-22kDa (reduced)
Positive Control: VEGF165 recombinant protein, Angiosarcoma Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-7 does not neutralize the bioactivity of VEGF. Recommended for: Note: USA: RUO; Int'l: RUO
Specifications
Vascular Endothelial Growth Factor (VEGF) Ab-7 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Angiogenesis | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin), Western Blot | |
VG1 | |
Unconjugated | |
Recombinant VEGF protein | |
RUO | |
Primary | |
Concentrated |