missing translation for 'onlineSavingsMsg'
Learn More

Vascular Endothelial Growth Factor (VEGF) Ab-7 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Vascular Endothelial Growth Factor (VEGF) Ab-7, Mouse Monoclonal Antibody.

Brand:  Epredia MS-1467-P0

Code : LD

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12639876

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

VEGF which is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.

Host Species: Mouse

Clone: VG1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant VEGF protein

Molecular Weight: 19-22kDa (reduced)

Positive Control: VEGF165 recombinant protein, Angiosarcoma

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-7 does not neutralize the bioactivity of VEGF.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Vascular Endothelial Growth Factor (VEGF) Ab-7
Monoclonal
200μg/mL
Mouse
100 μL
Angiogenesis
Human
IgG2a κ
Immunohistochemistry (Paraffin), Western Blot
VG1
Unconjugated
Recombinant VEGF protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers