Learn More
CD47 Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunofluorescence, immunoprecipitation and flow cytometry experiments with Epredia™ CD47 Ab-2, Mouse Monoclonal Antibody.
£103.02 - £579.00
Specifications
Antigen | CD47 Ab-2 |
---|---|
Clone | B6H12.2 |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12609536
|
Epredia
MS-1302-P1 |
200μg/mL | 500 μL |
£341.00
500µL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12649526
|
Epredia
MS-1302-P |
200μg/mL | 1 mL |
£547.00
1mL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12699526
|
Epredia
MS-1302-PABX |
1mg/mL | 200 μL |
£579.00
200µL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12689526
|
Epredia
MS-1302-P1ABX |
1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12679526
|
Epredia
MS-1302-P0 |
200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
CD47 antigen, also known as integrin associated-protein (IAP), is expressed on all hematopoietic cells, including leukocytes, platelets and erythrocytes. It is also expressed on epithelial cells, endothelial cells, fibroblasts and many tumor cell lines. CD47 may play a role as a signal transducer in the regulation of cation fluxes across cell membranes and in the chemotactic and adhesive interactions of leukocytes with endothelial cells.Host Species: Mouse
Clone: B6H12.2
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Ig domain
Immunogen: Intact CD47 purified from placenta
Molecular Weight: 47-52 kDa (reduced) and 45-50.5, 110 kDa (nonreduced)
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunofluorescence (Methanol fixed only)
- Immunoprecipitation (Native only)
- Flow Cytometry
- Blocks binding of SIRPα
- Inhibits in assays in which CD47-integrin association is required
Note:
USA: RUO; Int'l: RUO
Specifications
CD47 Ab-2 | |
Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Hematopoietic | |
Intact CD47 purified from placenta | |
Primary |
B6H12.2 | |
Monoclonal | |
Concentrated | |
RUO | |
Human | |
IgG2a κ |