Learn More
CD47 Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunofluorescence, immunoprecipitation and flow cytometry experiments with Epredia™ CD47 Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia MS-1302-P1ABX
Code : LD
Additional Details : Weight : 1.00000kg
Description
CD47 antigen, also known as integrin associated-protein (IAP), is expressed on all hematopoietic cells, including leukocytes, platelets and erythrocytes. It is also expressed on epithelial cells, endothelial cells, fibroblasts and many tumor cell lines. CD47 may play a role as a signal transducer in the regulation of cation fluxes across cell membranes and in the chemotactic and adhesive interactions of leukocytes with endothelial cells.Host Species: Mouse
Clone: B6H12.2
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Ig domain
Immunogen: Intact CD47 purified from placenta
Molecular Weight: 47-52 kDa (reduced) and 45-50.5, 110 kDa (nonreduced)
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunofluorescence (Methanol fixed only)
- Immunoprecipitation (Native only)
- Flow Cytometry
- Blocks binding of SIRPα
- Inhibits in assays in which CD47-integrin association is required
Note:
USA: RUO; Int'l: RUO
Specifications
CD47 Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Hematopoietic | |
Human | |
IgG2a κ |
Immunofluorescence, Immunoprecipitation, Western Blot | |
B6H12.2 | |
Unconjugated | |
Intact CD47 purified from placenta | |
RUO | |
Primary | |
Concentrated |