missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-1 Mouse, Clone: GAL1/1831, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | Galectin-1 |
---|---|
Clone | GAL1/1831 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960398
|
EnquireBio
3956-MSM1-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950398
|
EnquireBio
3956-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Galectin-1 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
3956 | |
Recombinant human Galectin-1 protein fragment (aa12-108) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
14kDa |
GAL1/1831 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_002296 | |
LGALS1 | |
IgG1 λ | |
Protein A/G Purified | |
Secreted (extracellular matrix) | |
HeLa, K562 or 293 cells. Prostate, Kidney, Placenta, Stomach, Skin, Spleen, Brain or Heart. |