missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-1 Mouse, Clone: GAL1/1831, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3956-MSM1-P0
Additional Details : Weight : 0.01000kg
Specifications
Galectin-1 | |
Secreted (extracellular matrix) | |
GAL1/1831 | |
Unconjugated | |
NP_002296 | |
Mouse | |
14kDa | |
Protein A/G Purified | |
RUO | |
3956 | |
IgG1 λ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
LGALS1 | |
Recombinant human Galectin-1 protein fragment (aa12-108) | |
HeLa, K562 or 293 cells. Prostate, Kidney, Placenta, Stomach, Skin, Spleen, Brain or Heart. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |