missing translation for 'onlineSavingsMsg'
Learn More

Galectin-1 Mouse, Clone: GAL1/1831, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 3956-MSM1-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960398

  • £395.00 / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Galectin-1
Secreted (extracellular matrix)
GAL1/1831
Unconjugated
NP_002296
Mouse
14kDa
Protein A/G Purified
RUO
3956
IgG1 λ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
LGALS1
Recombinant human Galectin-1 protein fragment (aa12-108)
HeLa, K562 or 293 cells. Prostate, Kidney, Placenta, Stomach, Skin, Spleen, Brain or Heart.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers