missing translation for 'onlineSavingsMsg'
Learn More

VACUUBRAND™ Centering Ring With NBR Sealing and Filter Insert

£34.40 - £71.75

Specifications

Quantity 1 Each
Type Centering Ring
View More Specs

Products 4
Product Code Brand For Use With Price Quantity & Availability  
Product Code Brand For Use With Price Quantity & Availability  
10187141
View Documents Promotion Details
VACUUBRAND™
20660160
For KF DN 10mm small flange
£34.40
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
 
10672862
View Documents Promotion Details
VACUUBRAND™
20660161
For KF DN 16mm small flange
£39.10
Each
View alternative products
Save Up to 
»
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
 
10731843
View Documents Promotion Details
VACUUBRAND™
20660163
For KF DN 40mm small flange
£71.75
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
 
10543562
View Documents Promotion Details
VACUUBRAND™
20660162
For KF DN 25mm small flange
£53.65
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

1 Each
Centering Ring
Videos
SDS
Documents

Documents

Product Certifications