Learn More
VACUUBRAND™ Centering Ring With NBR Sealing and Filter Insert
Brand:  VACUUBRAND™ 20660160
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10187141
  • £34.40 / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket
Specifications
For KF DN 10mm small flange
1 Each
Centering Ring