missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VACUUBRAND™ Centering Ring With NBR Sealing and Filter Insert
Brand: VACUUBRAND™ 20660161
32.20 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Code : A0
Additional Details : Weight : 0.50000kg
Specifications
For KF DN 16mm small flange | |
1 Each | |
Centering Ring |