Learn More
Hirschmann™ Amber Glass Stoppers
Light weight amber colored hollow glass stopper for the protection of UV-sensitive media.
£5.50 - £22.35
Specifications
For Use With (Equipment) | For tight sealing of standard ground vessels |
---|---|
Product Type Specs | Stopper |
Quantity | 1/Ea. |
Material | Glass |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
10261973
|
Hirschmann™
8902360 |
NS 10/19 |
£5.50
Each |
Log in to see stock. |
10526053
|
Hirschmann™
8902352 |
NS 7/16 |
£5.50
Each |
Log in to see stock. |
10097552
|
Hirschmann™
8902365 |
NS 12/21 |
£5.50
Each |
Log in to see stock. |
10556053
|
Hirschmann™
8902370 |
NS 14/23 |
£5.50
Each |
Log in to see stock. |
10097602
|
Hirschmann™
8902386 |
NS 29/32 |
£11.15
Each |
Log in to see stock. |
10252023
|
Hirschmann™
8902380 |
NS 24/29 |
£9.95
Each |
Log in to see stock. |
10189432
|
Hirschmann™
8902375 |
NS 19/26 |
£7.75
Each |
Log in to see stock. |
10417363
|
Hirschmann™
8902390 |
NS 34/35 |
£16.10
Each |
Log in to see stock. |
10057652
|
Hirschmann™
8902393 |
NS 45/40 |
£22.35
Each |
Log in to see stock. |
Description
- Acc. to DIN 12 252
Specifications
For tight sealing of standard ground vessels | |
Stopper | |
1/Ea. | |
Glass |