Learn More
Hirschmann™ Amber Glass Stoppers
Light weight amber colored hollow glass stopper for the protection of UV-sensitive media.
Brand:  Hirschmann™ 8902365
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10097552
  • £6.28 / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12 252
Specifications
Specifications
Stopper
Glass
NS 12/21
1/Ea.
For tight sealing of standard ground vessels