Learn More
Hirschmann™ Amber Glass Stoppers
Light weight amber colored hollow glass stopper for the protection of UV-sensitive media.
Brand:  Hirschmann™ 8902360
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10261973
  • £6.30 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12 252
Specifications
Specifications
Stopper
Glass
NS 10/19
1/Ea.
For tight sealing of standard ground vessels