Learn More
Hirschmann™ Amber Glass Stoppers
Light weight amber colored hollow glass stopper for the protection of UV-sensitive media.
Brand:  Hirschmann™ 8902393
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10057652
  • £25.60 / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12 252
Specifications
Specifications
Stopper
NS 45/40
Hexagonal
1 Each
DIN 12 252