Learn More
1,1-Dimethylbiguanide hydrochloride, Thermo Scientific™
1,1-Dimethylbiguanide hydrochloride (metformin hydrochloride, CAS # 115-70-4) is the salt of methylated biguanide, a compound exhibiting anti-hyperglycemic properties.
£31.65 - £229.00
Chemical Identifiers
| CAS | 1115-70-4 |
|---|---|
| Molecular Formula | C4H12ClN5 |
| Molecular Weight (g/mol) | 165.63 |
| MDL Number | MFCD00012582 |
| InChI Key | OETHQSJEHLVLGH-UHFFFAOYSA-N |
| Synonym | metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform |
| PubChem CID | 14219 |
| IUPAC Name | hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride |
| SMILES | [H+].[Cl-].CN(C)C(=N)N=C(N)N |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15485609
|
Thermo Scientific Alfa Aesar
J63361.06 |
5g |
£34.90 £31.65 / 5g Save £3.25 9% Off |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15495609
|
Thermo Scientific Alfa Aesar
J63361.14 |
25g |
£84.80 £76.65 / 25g Save £8.15 9% Off |
Estimated Shipment: 01-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15405619
|
Thermo Scientific Alfa Aesar
J63361.22 |
100g |
£229.00
100g |
Estimated Shipment: 07-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• 1,1-Dimethylbiguanide consists of two guanidines with methyl substitutions at position 1
• 1,1-Dimethylbiguanide hydrochloride inhibits complex I of the mitochondrial respiratory chain, increasing AMP and activating target genes
• 1,1-Dimethylbiguanide hydrochloride enhances insulin sensitivity and fatty acid oxidation in cells
• 1,1-Dimethylbiguanide hydrochloride may also have antineoplastic action through the mammalian target of rapamycin pathway
• It is suitable for the laboratory research of diabetes, obesity, and related metabolic disorders
• It is also useful for the laboratory study of mammalian target of rapamycin (mTOR) pathways and AMPK ratios within the cell
Chemical Identifiers
| C4H12ClN5 | |
| MFCD00012582 | |
| metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform | |
| hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride |
Specifications
| 1115-70-4 | |
| NH2C(=NH)NHC(=NH)N(CH3)2·HCl | |
| 14,5938 | |
| Soluble in water (50mg/ml),ethanol,and DMSO (50 mM). | |
| [H+].[Cl-].CN(C)C(=N)N=C(N)N | |
| 165.63 | |
| 165.62 |
| C4H12ClN5 | |
| MFCD00012582 | |
| metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform | |
| OETHQSJEHLVLGH-UHFFFAOYSA-N | |
| hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride | |
| 14219 | |
| 1,1-Dimethylbiguanide hydrochloride |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 214-230-6
missing translation for 'rtecsNumber' : DU1800000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.