missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylbiguanide hydrochloride

Product Code. 15495609
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495609 25 g 25g
15485609 5 g 5g
15405619 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15495609 Supplier Thermo Scientific Alfa Aesar Supplier No. J63361.14
  • ONLINE EXCLUSIVE PRICE
    £84.80
    £76.65 / 25g
    Save £8.15
    9% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An antidiabetic agent | CAS: 1115-70-4 | C4H12ClN5 | 165.63 g/mol

1,1-Dimethylbiguanide hydrochloride is used as an antidiabetic agent, used in polycystic ovary syndrome, non-alcoholic fatty liver disease and diabetes mellitus type 2. It may also reduce weight gain in people taking atypical antipsychotics, used in a study to investigate its role in enhancing the secretions of plasma active glucagon-like peptide-1 (GLP-1) levels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 1,1-Dimethylbiguanide consists of two guanidines with methyl substitutions at position 1

• 1,1-Dimethylbiguanide hydrochloride inhibits complex I of the mitochondrial respiratory chain, increasing AMP and activating target genes

• 1,1-Dimethylbiguanide hydrochloride enhances insulin sensitivity and fatty acid oxidation in cells

• 1,1-Dimethylbiguanide hydrochloride may also have antineoplastic action through the mammalian target of rapamycin pathway

• It is suitable for the laboratory research of diabetes, obesity, and related metabolic disorders

• It is also useful for the laboratory study of mammalian target of rapamycin (mTOR) pathways and AMPK ratios within the cell

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-70-4
Molecular Formula C4H12ClN5
Molecular Weight (g/mol) 165.63
MDL Number MFCD00012582
InChI Key OETHQSJEHLVLGH-UHFFFAOYSA-N
Synonym metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
PubChem CID 14219
IUPAC Name hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
SMILES [H+].[Cl-].CN(C)C(=N)N=C(N)N

Specifications

Melting Point 218°C to 226°C
Color White
Linear Formula NH2C(=NH)NHC(=NH)N(CH3)2·HCl
Quantity 25 g
Merck Index 14,5938
Solubility Information Soluble in water (50mg/ml),ethanol,and DMSO (50 mM).
Formula Weight 165.62
Physical Form Powder
Chemical Name or Material 1,1-Dimethylbiguanide hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.