missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylbiguanide hydrochloride, Thermo Scientific™

1,1-Dimethylbiguanide hydrochloride (metformin hydrochloride, CAS # 115-70-4) is the salt of methylated biguanide, a compound exhibiting anti-hyperglycemic properties.

Brand:  Thermo Scientific Alfa Aesar J63361.06

33.15 GBP valid until 2026-09-25
BEST PRICE on Promo! Use promo code "27966" to get your promotional price.


Quantity 5g

 View more versions of this product

Product Code. 15485609

  • ONLINE EXCLUSIVE PRICE
    £34.90
    £31.65 / 5g
    Save £3.25
    9% Off
View all promotions
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• 1,1-Dimethylbiguanide consists of two guanidines with methyl substitutions at position 1

• 1,1-Dimethylbiguanide hydrochloride inhibits complex I of the mitochondrial respiratory chain, increasing AMP and activating target genes

• 1,1-Dimethylbiguanide hydrochloride enhances insulin sensitivity and fatty acid oxidation in cells

• 1,1-Dimethylbiguanide hydrochloride may also have antineoplastic action through the mammalian target of rapamycin pathway

• It is suitable for the laboratory research of diabetes, obesity, and related metabolic disorders

• It is also useful for the laboratory study of mammalian target of rapamycin (mTOR) pathways and AMPK ratios within the cell

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

1115-70-4
165.63
OETHQSJEHLVLGH-UHFFFAOYSA-N
14219
[H+].[Cl-].CN(C)C(=N)N=C(N)N
C4H12ClN5
MFCD00012582
metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride

Specifications

1115-70-4
NH2C(=NH)NHC(=NH)N(CH3)2·HCl
5g
metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
OETHQSJEHLVLGH-UHFFFAOYSA-N
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
14219
1,1-Dimethylbiguanide hydrochloride
C4H12ClN5
MFCD00012582
14,5938
Soluble in water (50mg/ml),ethanol,and DMSO (50 mM).
[H+].[Cl-].CN(C)C(=N)N=C(N)N
165.63
165.62
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Promo
Expires: 2026-09-25
Curiosity Goes Beyond
Promo Code: 27966
Push the boundaries of discovery with exceptional savings across essential research workflows: Cell Culture and Analysis driven by our Gibco™ product portfolio, Gene Expression powered by Applied Biosystems™ solutions, and Protein Expression enabled by Invitrogen™ and Thermo Scientific™ products.

Applying Promo and Adding...

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.