missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylbiguanide hydrochloride, Thermo Scientific™

1,1-Dimethylbiguanide hydrochloride (metformin hydrochloride, CAS # 115-70-4) is the salt of methylated biguanide, a compound exhibiting anti-hyperglycemic properties.

Brand:  Thermo Scientific Alfa Aesar J63361.22


Quantity 100g

 View more versions of this product

Product Code. 15405619

  • £229.00 / 100g
 Restricted Item
Estimated Shipment: 07-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• 1,1-Dimethylbiguanide consists of two guanidines with methyl substitutions at position 1

• 1,1-Dimethylbiguanide hydrochloride inhibits complex I of the mitochondrial respiratory chain, increasing AMP and activating target genes

• 1,1-Dimethylbiguanide hydrochloride enhances insulin sensitivity and fatty acid oxidation in cells

• 1,1-Dimethylbiguanide hydrochloride may also have antineoplastic action through the mammalian target of rapamycin pathway

• It is suitable for the laboratory research of diabetes, obesity, and related metabolic disorders

• It is also useful for the laboratory study of mammalian target of rapamycin (mTOR) pathways and AMPK ratios within the cell

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

1115-70-4
165.63
OETHQSJEHLVLGH-UHFFFAOYSA-N
14219
[H+].[Cl-].CN(C)C(=N)N=C(N)N
C4H12ClN5
MFCD00012582
metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride

Specifications

1115-70-4
NH2C(=NH)NHC(=NH)N(CH3)2·HCl
100g
metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
OETHQSJEHLVLGH-UHFFFAOYSA-N
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
14219
1,1-Dimethylbiguanide hydrochloride
C4H12ClN5
MFCD00012582
14,5938
Soluble in water (50mg/ml),ethanol,and DMSO (50 mM).
[H+].[Cl-].CN(C)C(=N)N=C(N)N
165.63
165.62
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,1-Dimethylbiguanide hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Metformin hydrochloride

SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.