accessibility menu, dialog, popup

UserName

CAS RN 75-01-4

ClCH2

CAS RN 75-01-4

IUPAC Name: chloroethene
Synonyms: vinyl chloride vinylchlorid vinile (cloruro di) cloroetileno chlorure de vinyle trovidur vinyl chloride monomer VCM vinyl chlorine ccris 621
Molecular Weight (g/mol): 62.5
Molecular Formula: C2H3Cl
InChi Key: BZHJMEDXRYGGRV-UHFFFAOYSA-N
SMILES: ClC=C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Solvent or Matrix 
  • (4)
  • (1)
Concentration 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)