missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinylchloride in Methanol 100μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 75-01-4 |
| Physical Form | Liquid |
| Molecular Formula | C2H3Cl |
| Concentration | 100 μg/mL |
| InChI Key | BZHJMEDXRYGGRV-UHFFFAOYSA-N |
| Type | Organic Single-component Standard Solution |
| SMILES | C=CCl |
| IUPAC Name | chloroethene |
| Solvent or Matrix | Methanol |
| Show More |
Safety and Handling
Product Identifier
- vinyl chloride; chloroethylene 100ug/ml in methanol
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash hands, forearms andface thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contains : methanol, vinyl chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction