missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinyl chloride 100 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Vinyl chloride |
| CAS | 75-01-4 |
| Analyte or Component Names | Vinyl chloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C2 H3 Cl |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Ethene, chloro-, Ethylene, chloro- (8CI), Chloroethene, 1-Chloroethene, 1-Chloroethylene, Chloroethylene, F 1140, Monochloroethylene, VCM, Vinyl C monomer, Vinyl chloride, Vinyl chloride monomer |
| InChI Formula | InChI=1S/C2H3Cl/c1-2-3/h2H,1H2 |
| SMILES | ClC=C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction