missing translation for 'onlineSavingsMsg'
Learn More

Vinylchloride in Methanol 10μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12962974 Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 12962974

Brand: Fisher Chemical P857610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 75-01-4
Physical Form Liquid
Molecular Formula C2H3Cl
Concentration 10 μg/mL
InChI Key BZHJMEDXRYGGRV-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C=CCl
IUPAC Name chloroethene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 62.496
PubChem CID 6338
Quantity 10 mL
ChEBI CHEBI:28509
Chemical Name or Material Vinylchloride (C2H3CL)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vinylchloride [CAS:75-01-4] 10 ug/ml in Methanol (P857610)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.