accessibility menu, dialog, popup

UserName

CAS RN 66104-23-2

CH3N(R)SCH3(R)(R)HNCH3SOHOO

CAS RN 66104-23-2

IUPAC Name: (2R,4R,7R)-4-[(methylsulfanyl)methyl]-6-propyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene; methanesulfonic acid
Synonyms: permax pergolide mesilate pergolide monomesylate pergolide monomethanesulfonate
Molecular Weight (g/mol): 410.59
Molecular Formula: C20H30N2O3S2
InChi Key: UWCVGPLTGZWHGS-ZORIOUSZSA-N
SMILES: CS(O)(=O)=O.CCCN1C[C@H](CSC)C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results