missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pergolide Mesylate, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Pergolide mesilate |
| CAS | 66104-23-2 |
| Molecular Formula | C19 H26 N2 S . C H4 O3 S |
| Quantity | 100 mg |
| Synonym | Indolo[4,3-fg]quinoline','ergoline deriv., Celance, LY 127809, NSC 319773, Pergolide mesylate, Pergolide methanesulfonate, Permax |
| InChI Formula | InChI=1S/C19H26N2S.CH4O3S/c1-3-7-21-11-13(12-22-2)8-16-15-5-4-6-17-19(15)14(10-20-17)9-18(16)21;1-5(2,3)4/h4-6,10,13,16,18,20H,3,7-9,11-12H2,1-2H3;1H3,(H,2,3,4)/t13-,16-,18-;/m1./s1 |
| SMILES | CCCN1C[C@H](CSC)C[C@H]2[C@H]1Cc3c[nH]c4cccc2c34.CS(=O)(=O)O |
| IUPAC Name | (6aR,9R,10aR)-9-(methylsulfanylmethyl)-7-propyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline;methanesulfonic acid |
| Molecular Weight (g/mol) | 410.59 |
| Formula Weight | 410.1698 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction