missing translation for 'onlineSavingsMsg'
Learn More

Pergolide mesylate, MedChemExpress

Product Code. 16485818
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485818

Brand: Medchem Express HY13720A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pergolide mesylate (Pergolide methanesulfonate), an Ergoline derivative, is a potent and orally active dopamine D1 and D2 receptors agonist. Pergolide mesylate can be used for Parkinson's disease and hyperprolactinaemia research.

Pergolide mesylate (Pergolide methanesulfonate), an Ergoline derivative, is a potent and orally active dopamine D1 and D2 receptors agonist. Pergolide mesylate can be used for Parkinson's disease and hyperprolactinaemia research.

Specifications

Chemical Name or Material Pergolide mesylate
CAS 66104-23-2
Color White
Purity Grade Notes Research
Molecular Formula C20H30N2O3S2
Quantity 10 mg
Synonym Pergolide methanesulfonate LY127809
Solubility Information DMSO : ≥ 25 mg/mL (60.89 mM)
SMILES [H][C@@]1(N(CCC)C[C@H](CSC)C[C@@]12[H])CC3=CNC4=CC=CC2=C43.CS(=O)(O)=O
Molecular Weight (g/mol) 410.59
Formula Weight 410.59
Percent Purity 99.45%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.